MilliporeSigma
  • Reduced chemokine C-C motif ligand 1 expression may negatively regulate colorectal cancer progression at liver metastatic sites.
Reduced chemokine C-C motif ligand 1 expression may negatively regulate colorectal cancer progression at liver metastatic sites.
Journal of cellular and molecular medicine (2024-03-20)
Miku Iwata, Ryuma Haraguchi, Riko Kitazawa, Chihiro Ito, Kohei Ogawa, Yasutsugu Takada, Sohei Kitazawa
ABSTRACT
Colorectal cancer (CRC) liver metastasis, albeit a stage-IV disease, is completely curable by surgical resection in selected patients. In addressing the molecular basics of this phenomenon, differentially expressed genes at primary and liver metastatic sites were screened by RNA sequencing with the use of paraffin-embedded surgical specimens. Chemokine C-C motif ligand 1 (CCL1), a chemotactic factor for a ligand of the chemokine C-C motif receptor 8 (CCR8), was isolated as one of the differentially expressed genes. Histological analysis revealed that the number of CCL1-positive cells, mainly tumour associated macrophages (TAMs) located in the stroma of CRC, decreased significantly at liver metastatic sites, while the expression level of CCR8 on CRC remained unchanged. To explore the biological significance of the CCL1-CCR8 axis in CRC, CCR8-positive CRC cell line Colo320DM was used to assess the effect of the CCL1-CCR8 axis on major signalling pathways, epithelial mesenchymal transition induction and cell motility. Upon stimulation of recombinant CCL1 (rCCL1), phosphorylation of AKT was observed in Colo320DM cells; on the other hand, the corresponding significant increase in MMP-2 levels demonstrated by RT-qPCR was nullified by siRNA (siCCR8). In the scratch test, rCCL1 treatment significantly increased the motility of Colo320DM cells, which was similarly nullified by siCCR8. Thus, the activation of the CCL1-CCR8 axis is a positive regulator of CRC tumour progression. Reduced CCL1 expression of TAMs at liver metastatic sites may partly explain the unique slow tumour progression of CRC, thus providing for a grace period for radical resection of metastatic lesions.
Sigma-Aldrich
Anti-CCL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution