MilliporeSigma
  • Detection of TDP-43 seeding activity in the olfactory mucosa from patients with frontotemporal dementia.
Detection of TDP-43 seeding activity in the olfactory mucosa from patients with frontotemporal dementia.
Alzheimer's & dementia : the journal of the Alzheimer's Association (2023-11-01)
Elena Fontana, Matilde Bongianni, Alberto Benussi, Erika Bronzato, Carlo Scialo, Luca Sacchetto, Annachiara Cagnin, Santina Castriciano, Emanuele Buratti, Fabrizio Gardoni, Maria Italia, Alberto Schreiber, Chiara Ferracin, Michele Fiorini, Kathy L Newell, Laura Cracco, Holly J Garringer, Maria Paola Cecchini, Magdalini Polymenidou, Alessandro Padovani, Salvatore Monaco, Giuseppe Legname, Bernardino Ghetti, Barbara Borroni, Gianluigi Zanusso
ABSTRACT
We assessed TAR DNA-binding protein 43 (TDP-43) seeding activity and aggregates detection in olfactory mucosa of patients with frontotemporal lobar degeneration with TDP-43-immunoreactive pathology (FTLD-TDP) by TDP-43 seeding amplification assay (TDP43-SAA) and immunocytochemical analysis. The TDP43-SAA was optimized using frontal cortex samples from 16 post mortem cases with FTLD-TDP, FTLD with tau inclusions, and controls. Subsequently, olfactory mucosa samples were collected from 17 patients with FTLD-TDP, 15 healthy controls, and three patients carrying MAPT variants. TDP43-SAA discriminated with 100% accuracy post mortem cases presenting or lacking TDP-43 neuropathology. TDP-43 seeding activity was detectable in the olfactory mucosa, and 82.4% of patients with FTLD-TDP tested positive, whereas 86.7% of controls tested negative (P < 0.001). Two out of three patients with MAPT mutations tested negative. In TDP43-SAA positive samples, cytoplasmatic deposits of phosphorylated TDP-43 in the olfactory neural cells were detected. TDP-43 aggregates can be detectable in olfactory mucosa, suggesting that TDP43-SAA might be useful for identifying and monitoring FTLD-TDP in living patients.
Sigma-Aldrich
Monoclonal Anti-β-Tubulin IV antibody produced in mouse, clone ONS.1A6, ascites fluid
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-TDP-43 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution