MilliporeSigma
  • Reduced binding of apoE4 to complement factor H promotes amyloid-β oligomerization and neuroinflammation.
Reduced binding of apoE4 to complement factor H promotes amyloid-β oligomerization and neuroinflammation.
EMBO reports (2023-05-08)
Larisa Chernyaeva, Giorgio Ratti, Laura Teirilä, Satoshi Fudo, Uni Rankka, Anssi Pelkonen, Paula Korhonen, Katarzyna Leskinen, Salla Keskitalo, Kari Salokas, Christina Gkolfinopoulou, Katrina E Crompton, Matti Javanainen, Lotta Happonen, Markku Varjosalo, Tarja Malm, Ville Leinonen, Angeliki Chroni, Päivi Saavalainen, Seppo Meri, Tommi Kajander, Adam Jm Wollman, Eija Nissilä, Karita Haapasalo
ABSTRACT
The APOE4 variant of apolipoprotein E (apoE) is the most prevalent genetic risk allele associated with late-onset Alzheimer's disease (AD). ApoE interacts with complement regulator factor H (FH), but the role of this interaction in AD pathogenesis is unknown. Here we elucidate the mechanism by which isoform-specific binding of apoE to FH alters Aβ1-42-mediated neurotoxicity and clearance. Flow cytometry and transcriptomic analysis reveal that apoE and FH reduce binding of Aβ1-42 to complement receptor 3 (CR3) and subsequent phagocytosis by microglia which alters expression of genes involved in AD. Moreover, FH forms complement-resistant oligomers with apoE/Aβ1-42 complexes and the formation of these complexes is isoform specific with apoE2 and apoE3 showing higher affinity to FH than apoE4. These FH/apoE complexes reduce Aβ1-42 oligomerization and toxicity, and colocalize with complement activator C1q deposited on Aβ plaques in the brain. These findings provide an important mechanistic insight into AD pathogenesis and explain how the strongest genetic risk factor for AD predisposes for neuroinflammation in the early stages of the disease pathology.
Sigma-Aldrich
Anti-Factor H Goat pAb, liquid, Calbiochem®
Sigma-Aldrich
Anti-Amyloid β Antibody, clone W0-2, clone WO2, from mouse