MilliporeSigma
  • Nitric Oxide Affects Heme Oxygenase-1, Hepcidin, and Transferrin Receptor Expression in the Placenta.
Nitric Oxide Affects Heme Oxygenase-1, Hepcidin, and Transferrin Receptor Expression in the Placenta.
International journal of molecular sciences (2023-03-30)
Patricia Principe, George T Mukosera, Nikia Gray-Hutto, Ashra Tugung, Ciprian P Gheorghe, Arlin B Blood
ABSTRACT
Nitric oxide (NO) is a gasotransmitter that avidly binds both free and heme-bound iron, forming relatively stable iron nitrosyl compounds (FeNOs). We have previously demonstrated that FeNOs are present in the human placenta and are elevated in preeclampsia and intrauterine growth restriction. The ability of NO to sequester iron raises the possibility of the NO-mediated disruption of iron homeostasis in the placenta. In this work, we tested whether exposure of placental syncytiotrophoblasts or villous tissue explants to sub-cytotoxic concentrations of NO would elicit the formation of FeNOs. Furthermore, we measured changes in the mRNA and protein expression levels of key iron regulatory genes in response to NO exposure. Ozone-based chemiluminescence was used to measure concentrations of NO and its metabolites. Our results showed a significant increase in FeNO levels in placental cells and explants treated with NO (p < 0.0001). The mRNA and protein levels of HO-1 were significantly increased in both cultured syncytiotrophoblasts and villous tissue explants (p < 0.01), and the mRNA levels of hepcidin and transferrin receptor were significantly increased in culture syncytiotrophoblasts and villous tissue explants, respectively, (p < 0.01), while no changes were seen in the expression levels of divalent metal transporter-1 or ferroportin. These results suggest a potential role for NO in iron homeostasis in the human placenta and could be relevant for disorders of pregnancy such as fetal growth restriction and preeclampsia.
Sigma-Aldrich
Monoclonal Anti-HAMP, (C-terminal) antibody produced in mouse, clone 1F9, purified immunoglobulin, buffered aqueous solution