MilliporeSigma
  • A comprehensive synthetic library of poly-N-acetyl glucosamines enabled vaccine against lethal challenges of Staphylococcus aureus.
A comprehensive synthetic library of poly-N-acetyl glucosamines enabled vaccine against lethal challenges of Staphylococcus aureus.
Nature communications (2024-04-25)
Zibin Tan, Weizhun Yang, Nicholas A O'Brien, Xingling Pan, Sherif Ramadan, Terence Marsh, Neal Hammer, Colette Cywes-Bentley, Mariana Vinacur, Gerald B Pier, Jeffrey C Gildersleeve, Xuefei Huang
ABSTRACT
Poly-β-(1-6)-N-acetylglucosamine (PNAG) is an important vaccine target, expressed on many pathogens. A critical hurdle in developing PNAG based vaccine is that the impacts of the number and the position of free amine vs N-acetylation on its antigenicity are not well understood. In this work, a divergent strategy is developed to synthesize a comprehensive library of 32 PNAG pentasaccharides. This library enables the identification of PNAG sequences with specific patterns of free amines as epitopes for vaccines against Staphylococcus aureus (S. aureus), an important human pathogen. Active vaccination with the conjugate of discovered PNAG epitope with mutant bacteriophage Qβ as a vaccine carrier as well as passive vaccination with diluted rabbit antisera provides mice with near complete protection against infections by S. aureus including methicillin-resistant S. aureus (MRSA). Thus, the comprehensive PNAG pentasaccharide library is an exciting tool to empower the design of next generation vaccines.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) Alkaline Phosphatase conjugate, 1.0 mg/mL, Chemicon®
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) Alkaline Phosphatase conjugate, 1 mg/mL, Chemicon®