MilliporeSigma
  • Uracil- and Pyridine-Containing HDAC Inhibitors Displayed Cytotoxicity in Colorectal and Glioblastoma Cancer Stem Cells.
Uracil- and Pyridine-Containing HDAC Inhibitors Displayed Cytotoxicity in Colorectal and Glioblastoma Cancer Stem Cells.
ChemMedChem (2024-03-26)
Francesco Fiorentino, Emanuele Fabbrizi, Alessia Raucci, Beatrice Noce, Rossella Fioravanti, Sergio Valente, Chantal Paolini, Ruggero De Maria, Christian Steinkühler, Paola Gallinari, Dante Rotili, Antonello Mai
ABSTRACT
Cancer stem cells (CSCs) are a niche of highly tumorigenic cells featuring self-renewal, activation of pluripotency genes, multidrug resistance, and ability to cause cancer relapse. Seven HDACi (1-7), showing either hydroxamate or 2'-aminoanilide function, were tested in colorectal cancer (CRC) and glioblastoma multiforme (GBM) CSCs to determine their effects on cell proliferation, H3 acetylation levels and in-cell HDAC activity. Two uracil-based hydroxamates, 5 and 6, which differ in substitution at C5 and C6 positions of the pyrimidine ring, exhibited the greatest cytotoxicity in GBM (5) and CRC (6) CSCs, followed by the pyridine-hydroxamate 2, with 2- to 6-fold higher potency than the positive control SAHA. Finally, increased H3 acetylation as well as HDAC inhibition directly in cells by selected 2'-aminoanilide 4 and hydroxamate 5 confirmed target engagement. Further investigation will be conducted into the broad-spectrum anticancer properties of the most potent derivatives and their effects in combination with approved, conventional anticancer drugs.
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
1 Estimated to ship on March 09, 2025
CHF 34.30
1 g
1 Estimated to ship on March 09, 2025
CHF 137.00
100 g
1 Estimated to ship on March 09, 2025
CHF 1,230.00
500 g
1 Estimated to ship on March 09, 2025
CHF 2,140.00
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
1 Estimated to ship on March 09, 2025
CHF 34.30
1 g
1 Estimated to ship on March 09, 2025
CHF 137.00
100 g
1 Estimated to ship on March 09, 2025
CHF 1,230.00
500 g
1 Estimated to ship on March 09, 2025
CHF 2,140.00