MilliporeSigma
  • Memory B cell responses induced by pneumococcal conjugate vaccine schedules with fewer doses: analysis of a randomised-controlled trial in Viet Nam.
Memory B cell responses induced by pneumococcal conjugate vaccine schedules with fewer doses: analysis of a randomised-controlled trial in Viet Nam.
Nature communications (2024-08-14)
Darren Suryawijaya Ong, Thanh V Phan, Beth Temple, Zheng Quan Toh, Cattram Duong Nguyen, Kien Vientrung, Hoang Van Anh Nguyen, Vo Thi Trang Dai, Kathryn Bright, Hau Phuc Tran, Rachel Ann Higgins, Yin Bun Cheung, Thuong Vu Nguyen, Kim Mulholland, Paul Vincent Licciardi
ABSTRACT
The use of pneumococcal conjugate vaccine (PCV) schedules with fewer doses are being considered to reduce costs and improve access, particularly in low- and middle-income countries. While several studies have assessed their immunogenicity, there are limited data on their potential for long-term immune protection, as assessed by pneumococcal serotype-specific memory B cell (Bmem) responses. This current study reports secondary outcome data that aims to compare Bmem responses following reduced-dose (0 + 1 and 1 + 1) schedules of PCV10 and PCV13 in Vietnamese infants from our randomised-controlled trial (trial registration number NCT03098628). Following vaccination at 12 months of age, Bmem levels for most serotypes peaked seven days post-vaccination and were higher in magnitude for the 1 + 1 than 0 + 1 schedules and for PCV13 than PCV10. Furthermore, Bmem did not wane as rapidly as IgG levels by 24 months of age. Further studies are needed to assess the use of Bmem as markers of long-term protection against pneumococcal carriage and disease, which is crucial to generate data for immunisation program decision-making.