MilliporeSigma
Immunostaining for Epigenetic Modifications in Fagopyrum Calli.
Methods in molecular biology (Clifton, N.J.) (2024-03-27)
Alicja Tomasiak, Katarzyna Sala, Agnieszka Braszewska
ABSTRACT
Immunostaining is a well-established technique for identifying specific proteins in tissue samples with specific antibodies to identify a single target protein. It is commonly used in research and provides information about cellular localization and protein expression levels. This chapter describes a detailed protocol for immunostaining fixed Fagopyrum calli embedded in Steedman's wax using nine antibodies raised against histone H3 and H4 methylation and acetylation on several lysines and DNA methylation.
Sigma-Aldrich
Anti-acetyl-Histone H4 (Lys12) Antibody, rabbit monoclonal, culture supernatant, from rabbit
Sigma-Aldrich
Anti-acetyl-Histone H4 (Lys5) Antibody, rabbit monoclonal, culture supernatant, Upstate®
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, clone AW30, rabbit monoclonal, culture supernatant, clone AW30, Upstate®
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, clone MC315, rabbit monoclonal, culture supernatant, clone MC315, Upstate®