MilliporeSigma
  • Fructose vs. glucose: modulating stem cell growth and function through sugar supplementation.
Fructose vs. glucose: modulating stem cell growth and function through sugar supplementation.
FEBS open bio (2024-06-26)
Salaheldeen Elsaid, Xiangdong Wu, Sui Seng Tee
ABSTRACT
In multicellular organisms, stem cells are impacted by microenvironmental resources such as nutrient availability and oxygen tension for their survival, growth, and differentiation. However, the accessibility of these resources in the pericellular environment greatly varies from organ to organ. This divergence in resource availability leads to variations in the potency and differentiation potential of stem cells. This study aimed to explore the distinct effects of glucose and fructose, as well as different oxygen tensions, on the growth dynamics, cytokine production, and differentiation of stem cells. We showed that replacing glucose with fructose subjected stem cells to stress, resulting in increased Hif1α expression and stability, which in turn led to a reduction in cell proliferation, and alterations in cytokine production. However, fructose failed to induce differentiation of human mesenchymal stem cells (hMSCs) as well as mouse fibroblasts into mature adipocytes compared to glucose, despite the upregulation of key markers of adipogenesis, including C/EBPβ, and PPARγ. Conversely, we showed that fructose induced undifferentiated mouse fibroblasts to release cytokines associated with senescence, including IL1α1, IL6, IL8, MCP1, and TNF1α, suggesting that these cells were undergoing lipolysis. Taken together, our results suggest that altering the culture conditions through changes in hexose levels and oxygen tension places considerable stress on stem cells. Additional research is required to further characterize the mechanisms governing stem cell response to their microenvironments.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, A cell-permeable, non-specific inhibitor of cAMP and cGMP phosphodiesterases (IC₅₀ = 2-50 µM).
Sigma-Aldrich
Indomethacin, A non-steroidal anti-inflammatory, cell permeable, antipyretic agent.
Sigma-Aldrich
Anti-KHK antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution