MilliporeSigma
  • Protective effect of colchicine on albumin glycation and cellular oxidative stress: Insights into diabetic cardiomyopathy.
Protective effect of colchicine on albumin glycation and cellular oxidative stress: Insights into diabetic cardiomyopathy.
Journal of biochemical and molecular toxicology (2024-02-19)
Muskan Rajkumar Thakur, Rashmi Santosh Tupe
ABSTRACT
The present work elucidates the role of colchicine (COL) on albumin glycation and cellular oxidative stress in diabetic cardiomyopathy (DCM). Human serum albumin (HSA) was glycated with methylglyoxal in the presence of COL (2.5, 3.75, and 5 µM), whereas positive and negative control samples were maintained separately. The effects of COL on HSA glycation, structural and functional modifications in glycated HSA were analyzed using different spectroscopical and fluorescence techniques. Increased fructosamine, carbonyl, and pentosidine formation in glycated HSA samples were inhibited in the presence of COL. Structural conformation of HSA and glycated HSA samples was examined by field emission scanning electron microscopy, circular dichroism, Fourier transform infrared, and proton nuclear magnetic resonance analyses, where COL maintained both secondary and tertiary structures of HSA against glycation. Functional marker assays included ABTS•+ radical scavenging and total antioxidant activities, advanced oxidative protein product formation, and turbidimetry, which showed preserved functional properties of glycated HSA in COL-containing samples. Afterward, rat cardiomyoblast (H9c2 cell line) was treated with glycated HSA-COL complex (400 μg/mL) for examining various cellular antioxidants (nitric oxide, catalase, superoxide dismutase, and glutathione) and detoxification enzymes (aldose reductase, glyoxalase I, and II) levels. All three concentrations of COL exhibited effective anti-glycation properties, enhanced cellular antioxidant levels, and detoxification enzyme activities. The report comprehensively analyzes the potential anti-glycation and properties of COL during its initial assessment.
Sigma-Aldrich
RAGE Antagonist, FPS-ZM1, RAGE Antagonist, FPS-ZM1, is a blood-brain-barrier permeant blocker of RAGE V domain-mediated ligand binding (Ki = 25, 148, & 230 nM, respectively, against Aβ40, HMGB1 & S100B, binding to sRAGE).