MilliporeSigma
  • Serpine1 mRNA confers mesenchymal characteristics to the cell and promotes CD8+ T cells exclusion from colon adenocarcinomas.
Serpine1 mRNA confers mesenchymal characteristics to the cell and promotes CD8+ T cells exclusion from colon adenocarcinomas.
Cell death discovery (2024-03-07)
Salvador Polo-Generelo, Cristina Rodríguez-Mateo, Belén Torres, José Pintor-Tortolero, José A Guerrero-Martínez, Julian König, Jesús Vázquez, Elena Bonzón-Kulichenco, Javier Padillo-Ruiz, Fernando de la Portilla, José C Reyes, José A Pintor-Toro
ABSTRACT
Serine protease inhibitor clade E member 1 (SERPINE1) inhibits extracellular matrix proteolysis and cell detachment. However, SERPINE1 expression also promotes tumor progression and plays a crucial role in metastasis. Here, we solve this apparent paradox and report that Serpine1 mRNA per se, independent of its protein-coding function, confers mesenchymal properties to the cell, promoting migration, invasiveness, and resistance to anoikis and increasing glycolytic activity by sequestering miRNAs. Expression of Serpine1 mRNA upregulates the expression of the TRA2B splicing factor without affecting its mRNA levels. Through transcriptional profiling, we found that Serpine1 mRNA expression downregulates through TRA2B the expression of genes involved in the immune response. Analysis of human colon tumor samples showed an inverse correlation between SERPINE1 mRNA expression and CD8+ T cell infiltration, unveiling the potential value of SERPINE1 mRNA as a promising therapeutic target for colon tumors.
Sigma-Aldrich
Anti-CD8A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution