MilliporeSigma
  • The deubiquitinase cofactor UAF1 interacts with USP1 and plays an essential role in spermiogenesis.
The deubiquitinase cofactor UAF1 interacts with USP1 and plays an essential role in spermiogenesis.
iScience (2024-04-09)
Ziqi Wang, Tongtong Li, Dongkai Liu, Mengjing Li, Shangming Liu, Xiaochen Yu, Hanzhen Li, Hui Song, Wei Zhao, Zhaojian Liu, Xiangfeng Chen, Gang Lu, Zi-Jiang Chen, Tao Huang, Hongbin Liu
ABSTRACT
Spermiogenesis defines the final phase of male germ cell differentiation. While multiple deubiquitinating enzymes have been linked to spermiogenesis, the impacts of deubiquitination on spermiogenesis remain poorly characterized. Here, we investigated the function of UAF1 in mouse spermiogenesis. We selectively deleted Uaf1 in premeiotic germ cells using the Stra8-Cre knock-in mouse strain (Uaf1 sKO), and found that Uaf1 is essential for spermiogenesis and male fertility. Further, UAF1 interacts and colocalizes with USP1 in the testes. Conditional knockout of Uaf1 in testes results in disturbed protein levels and localization of USP1, suggesting that UAF1 regulates spermiogenesis through the function of the deubiquitinating enzyme USP1. Using tandem mass tag-based proteomics, we identified that conditional knockout of Uaf1 in the testes results in reduced levels of proteins that are essential for spermiogenesis. Thus, we conclude that the UAF1/USP1 deubiquitinase complex is essential for normal spermiogenesis by regulating the levels of spermiogenesis-related proteins.