MilliporeSigma
  • Organelle resolved proteomics uncovers PLA2R1 as a novel cell surface marker required for chordoma growth.
Organelle resolved proteomics uncovers PLA2R1 as a novel cell surface marker required for chordoma growth.
Acta neuropathologica communications (2024-03-08)
Shahbaz Khan, Jeffrey A Zuccato, Vladimir Ignatchenko, Olivia Singh, Meinusha Govindarajan, Matthew Waas, Salvador Mejia-Guerrero, Andrew Gao, Gelareh Zadeh, Thomas Kislinger
ABSTRACT
Chordomas are clinically aggressive tumors with a high rate of disease progression despite maximal therapy. Given the limited therapeutic options available, there remains an urgent need for the development of novel therapies to improve clinical outcomes. Cell surface proteins are attractive therapeutic targets yet are challenging to profile with common methods. Four chordoma cell lines were analyzed by quantitative proteomics using a differential ultracentrifugation organellar fractionation approach. A subtractive proteomics strategy was applied to select proteins that are plasma membrane enriched. Systematic data integration prioritized PLA2R1 (secretory phospholipase A2 receptor-PLA2R1) as a chordoma-enriched surface protein. The expression profile of PLA2R1 was validated across chordoma cell lines, patient surgical tissue samples, and normal tissue lysates via immunoblotting. PLA2R1 expression was further validated by immunohistochemical analysis in a richly annotated cohort of 25-patient tissues. Immunohistochemistry analysis revealed that elevated expression of PLA2R1 is correlated with poor prognosis. Using siRNA- and CRISPR/Cas9-mediated knockdown of PLA2R1, we demonstrated significant inhibition of 2D, 3D and in vivo chordoma growth. PLA2R1 depletion resulted in cell cycle defects and metabolic rewiring via the MAPK signaling pathway, suggesting that PLA2R1 plays an essential role in chordoma biology. We have characterized the proteome of four chordoma cell lines and uncovered PLA2R1 as a novel cell-surface protein required for chordoma cell survival and association with patient outcome.
Sigma-Aldrich
Anti-PLA2R Antibody, clone 5F5.1, clone 5F5.1, from mouse
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder