MilliporeSigma
Inferring regulators of cell identity in the human adult pancreas.
NAR genomics and bioinformatics (2023-07-12)
Lotte Vanheer, Federica Fantuzzi, San Kit To, Andrea Schiavo, Matthias Van Haele, Tessa Ostyn, Tine Haesen, Xiaoyan Yi, Adrian Janiszewski, Joel Chappell, Adrien Rihoux, Toshiaki Sawatani, Tania Roskams, Francois Pattou, Julie Kerr-Conte, Miriam Cnop, Vincent Pasque
ABSTRACT
Cellular identity during development is under the control of transcription factors that form gene regulatory networks. However, the transcription factors and gene regulatory networks underlying cellular identity in the human adult pancreas remain largely unexplored. Here, we integrate multiple single-cell RNA-sequencing datasets of the human adult pancreas, totaling 7393 cells, and comprehensively reconstruct gene regulatory networks. We show that a network of 142 transcription factors forms distinct regulatory modules that characterize pancreatic cell types. We present evidence that our approach identifies regulators of cell identity and cell states in the human adult pancreas. We predict that HEYL, BHLHE41 and JUND are active in acinar, beta and alpha cells, respectively, and show that these proteins are present in the human adult pancreas as well as in human induced pluripotent stem cell (hiPSC)-derived islet cells. Using single-cell transcriptomics, we found that JUND represses beta cell genes in hiPSC-alpha cells. BHLHE41 depletion induced apoptosis in primary pancreatic islets. The comprehensive gene regulatory network atlas can be explored interactively online. We anticipate our analysis to be the starting point for a more sophisticated dissection of how transcription factors regulate cell identity and cell states in the human adult pancreas.
Sigma-Aldrich
Monoclonal Anti-Glucagon antibody produced in mouse, clone K79bB10, ascites fluid
Sigma-Aldrich
Anti-Achaete Scute homolog 2 Antibody, clone 7E2, clone 7E2, from mouse
Sigma-Aldrich
Anti-BHLHE41 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody