MilliporeSigma
  • ER stress elicits non-canonical CASP8 (caspase 8) activation on autophagosomal membranes to induce apoptosis.
ER stress elicits non-canonical CASP8 (caspase 8) activation on autophagosomal membranes to induce apoptosis.
Autophagy (2023-09-21)
Tatsuya Hattori, Kevin A Fundora, Kouta Hamamoto, David M Opozda, Xinwen Liang, Xiaoming Liu, Jiawen Zhang, Yasin Uzun, Yoshinori Takahashi, Hong-Gang Wang
ABSTRACT
The VPS37A gene encodes a subunit of the endosomal sorting complex required for transport (ESCRT)-I complex that is frequently lost in a wide variety of human solid cancers. We have previously demonstrated the role of VPS37A in directing the ESCRT membrane scission machinery to seal the phagophore for autophagosome completion. Here, we report that VPS37A-deficient cells exhibit an accumulation of the apoptotic initiator CASP8 (caspase 8) on the phagophore and are primed to undergo rapid apoptosis through the intracellular death-inducing signaling complex (iDISC)-mediated CASP8 activation upon exposure to endoplasmic reticulum (ER) stress. Using CRISPR-Cas9 gene editing and comparative transcriptome analysis, we identified the ATF4-mediated stress response pathway as a crucial mediator to elicit iDISC-mediated apoptosis following the inhibition of autophagosome closure. Notably, ATF4-mediated iDISC activation occurred independently of the death receptor TNFRSF10B/DR5 upregulation but required the pro-apoptotic transcriptional factor DDIT3/CHOP to enhance the mitochondrial amplification pathway for full-activation of CASP8 in VPS37A-deficient cells stimulated with ER stress inducers. Our analysis also revealed the upregulation of NFKB/NF-kB signaling as a potential mechanism responsible for restraining iDISC activation and promoting cell survival upon VPS37A depletion. These findings have important implications for the future development of new strategies to treat human cancers, especially those with VPS37A loss.Abbreviations: ATG: autophagy related; BMS: BMS-345541; CASP: caspase; CHMP: charged multivesicular body protein; DKO: double knockout; Dox: doxycycline; ER: endoplasmic reticulum; ESCRT: endosomal sorting complex required for transport; gRNA: guide RNA; GSEA: gene set enrichment analysis; GSK157: GSK2656157; iDISC: intracellular death-inducing signaling complex; IKK: inhibitor of NFKB kinase; IPA: ingenuity pathway analysis; KO: knockout; MAP1LC3/LC3: microtubule associated protein 1 light chain 3; NFKB/NF-kB: nuclear factor kappa B; OZ: 5Z-7-oxozeaenol; RNA-seq: RNA sequencing; UPR: unfolded protein response; TFT: transcription factor target; THG: thapsigargin; TUN: tunicamycin; VPS: vacuolar protein sorting.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
BMS-345541, ≥98% (HPLC), powder