MilliporeSigma
  • Differential use of importin-α isoforms governs cell tropism and host adaptation of influenza virus.
Differential use of importin-α isoforms governs cell tropism and host adaptation of influenza virus.
Nature communications (2011-01-20)
Gülsah Gabriel, Karin Klingel, Anna Otte, Swantje Thiele, Ben Hudjetz, Gökhan Arman-Kalcek, Martina Sauter, Tatiana Shmidt, Franziska Rother, Sigrid Baumgarte, Björn Keiner, Enno Hartmann, Michael Bader, George G Brownlee, Ervin Fodor, Hans-Dieter Klenk
ABSTRACT
Influenza A viruses are a threat to humans due to their ability to cross species barriers, as illustrated by the 2009 H1N1v pandemic and sporadic H5N1 transmissions. Interspecies transmission requires adaptation of the viral polymerase to importin-α, a cellular protein that mediates transport into the nucleus where transcription and replication of the viral genome takes place. In this study, we analysed replication, host specificity and pathogenicity of avian and mammalian influenza viruses, in importin-α-silenced cells and importin-α-knockout mice, to understand the role of individual importin-α isoforms in adaptation. For efficient virus replication, the polymerase subunit PB2 and the nucleoprotein (NP) of avian viruses required importin-α3, whereas PB2 and NP of mammalian viruses showed importin-α7 specificity. H1N1v replication depended on both, importin-α3 and -α7, suggesting ongoing adaptation of this virus. Thus, differences in importin-α specificity are determinants of host range underlining the importance of the nuclear envelope in interspecies transmission.
Product Description
Millipore
FLAG® Peptide, lyophilized powder
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution