MilliporeSigma
Multimaterial and multiscale scaffold for engineering enthesis organ.
International journal of bioprinting (2023-07-17)
Simone Micalizzi, Lara Russo, Chiara Giacomelli, Francesca Montemurro, Carmelo de Maria, Martina Nencioni, Laura Marchetti, Maria Letizia Trincavelli, Giovanni Vozzi
ABSTRACT
Tendon and ligament injuries are relevant clinical problems in modern society, and the current medical approaches do not guarantee complete recovery of the physiological functionalities. Moreover, they present a non-negligible failure rate after surgery. Failures often occur at the enthesis, which is the area of tendons and ligaments insertion to bones. This area is highly anisotropic and composed of four distinct zones: tendon or ligament, non-mineralized fibrocartilage, mineralized fibrocartilage, and bone. The organization of these regions provides a gradient in mechanical properties, biochemical composition, cellular phenotype, and extracellular matrix organization. Tissue engineering represents an alternative to traditional medical approaches. This work presents a novel biofabrication approach for engineering the enthesis. Gradient-based scaffolds were fabricated by exploiting the combination of electrospinning and three-dimensional (3D) bioprinting technologies. Studies were conducted to evaluate scaffold biocompatibility by seeding bone marrow-derived mesenchymal stem cells (BM-MSCs). Then, the scaffold's ability to promote cellular adhesion, growth, proliferation, and differentiation in both tenogenic and osteogenic phenotypes was evaluated. Fabricated scaffolds were also morphologically and mechanically characterized, showing optimal properties comparable to literature data. The versatility and potentiality of this novel biofabrication approach were demonstrated by fabricating clinical-size 3D enthesis scaffolds. The mechanical characterization highlighted their behavior during a tensile test was comparable to tendons and ligaments in vivo.
Sigma-Aldrich
Anti-THY1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®