MilliporeSigma
  • Potent induction of trained immunity by Saccharomyces cerevisiae β-glucans.
Potent induction of trained immunity by Saccharomyces cerevisiae β-glucans.
Frontiers in immunology (2024-02-28)
Patricia Vuscan, Brenda Kischkel, Aikaterini Hatzioannou, Efrosyni Markaki, Andrei Sarlea, Maria Tintoré, Jordi Cuñé, Panayotis Verginis, Carlos de Lecea, Triantafyllos Chavakis, Leo A B Joosten, Mihai G Netea
ABSTRACT
Candida albicans cell wall component β-glucan has been extensively studied for its ability to induce epigenetic and functional reprogramming of innate immune cells, a process termed trained immunity. We show that a high-complexity blend of two individual β-glucans from Saccharomyces cerevisiae possesses strong bioactivity, resulting in an enhanced trained innate immune response by human primary monocytes. The training required the Dectin-1/CR3, TLR4, and MMR receptors, as well as the Raf-1, Syk, and PI3K downstream signaling molecules. By activating multiple receptors and downstream signaling pathways, the components of this β-glucan preparation are able to act synergistically, causing a robust secondary response upon an unrelated challenge. In in-vivo murine models of melanoma and bladder cell carcinoma, pre-treatment of mice with the β-glucan preparation led to a significant reduction in tumor growth. These insights may aid in the development of future therapies based on β-glucan structures that induce an effective trained immunity response.
Sigma-Aldrich
Syk Inhibitor, The Spleen Tyrosine Kinase Inhibitor, also referenced under CAS 622387-85-3, controls the biological activity of Spleen Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.