MilliporeSigma
  • Renal Medullary Overexpression of Sphingosine-1-Phosphate Receptor 1 Transgene Attenuates Deoxycorticosterone Acetate (DOCA)-Salt Hypertension.
Renal Medullary Overexpression of Sphingosine-1-Phosphate Receptor 1 Transgene Attenuates Deoxycorticosterone Acetate (DOCA)-Salt Hypertension.
American journal of hypertension (2023-05-12)
Gaizun Hu, Dengpiao Xie, Chaoling Chen, Weili Wang, Pin-Lan Li, Joseph K Ritter, Ningjun Li
ABSTRACT
Our previous studies showed that renal medullary sphingosine-1-phosphate receptor 1 (S1PR1) mediated sodium excretion, high salt intake increased S1PR1 level, deoxycorticosterone acetate (DOCA) blocked high salt-induced S1PR1 in the renal medulla, and that conditional knockout of S1PR1 in the collecting duct aggravated DOCA-salt hypertension. The present study tested the hypothesis that overexpression of S1PR1 transgene in the renal medulla attenuates the sodium retention and hypertension in DOCA-salt mouse model. Male C57BL/6J mice received renal medullary transfection of control or S1PR1-expressing plasmids and then DOCA-salt treatment. Renal sodium excretion and arterial pressure were compared between control and S1PR1-overexpressed mice in response to high salt loading or pressure natriuresis. S1PR1-transfected mice showed significantly enhanced urinary sodium excretion in response to acute sodium loading (0.93 ± 0.27 in control vs. 4.72 ± 1.12 µmol/min/gKW in S1PR1-overexpressed mice, P < 0.05) and the pressure natriuresis (3.58 ± 1.77 vs. 9.52 ± 1.38, P < 0.05), less positive sodium balance in response to chronic high-salt intake (3.05 ± 0.39 vs. 1.65 ± 0.39 mmol/72 hr, P < 0.05), and consequently, the attenuation of DOCA-salt hypertension (134.2 ± 6.79 vs. 109.8 ± 3.54 mm Hg, P < 0.05). The αENaC protein amount in the renal medulla was not changed, however, the βENaC was significantly decreased and the γENaC was significantly increased in S1PR1-overexpressed mice. The immunostaining showed apical membrane translocation of γENaC, while no change of αENaC and βENaC in control mice, and that the apical membrane translocation of γENaC was blocked in S1PR1-treasffected mice. These results suggested that activation of S1PR1 in the renal medulla attenuates DOCA-induced sodium retention and salt-sensitive hypertension associated with inhibition of ENaC.
Sigma-Aldrich
Anti-Epithelial Sodium Channel-α Antibody, Chemicon®, from rabbit