MilliporeSigma
Aberrant cortex contractions impact mammalian oocyte quality.
Developmental cell (2024-02-23)
Elvira Nikalayevich, Gaëlle Letort, Ghislain de Labbey, Elena Todisco, Anastasia Shihabi, Hervé Turlier, Raphaël Voituriez, Mohamed Yahiatene, Xavier Pollet-Villard, Metello Innocenti, Melina Schuh, Marie-Emilie Terret, Marie-Hélène Verlhac
ABSTRACT
The cortex controls cell shape. In mouse oocytes, the cortex thickens in an Arp2/3-complex-dependent manner, ensuring chromosome positioning and segregation. Surprisingly, we identify that mouse oocytes lacking the Arp2/3 complex undergo cortical actin remodeling upon division, followed by cortical contractions that are unprecedented in mammalian oocytes. Using genetics, imaging, and machine learning, we show that these contractions stir the cytoplasm, resulting in impaired organelle organization and activity. Oocyte capacity to avoid polyspermy is impacted, leading to a reduced female fertility. We could diminish contractions and rescue cytoplasmic anomalies. Similar contractions were observed in human oocytes collected as byproducts during IVF (in vitro fertilization) procedures. These contractions correlate with increased cytoplasmic motion, but not with defects in spindle assembly or aneuploidy in mice or humans. Our study highlights a multiscale effect connecting cortical F-actin, contractions, and cytoplasmic organization and affecting oocyte quality, with implications for female fertility.
Sigma-Aldrich
ML-7, powder
Sigma-Aldrich
Monastrol, ≥98% (HPLC), solid
Rabbit IgG HRP Linked Whole Ab, Cytiva NA934-1ML
Mouse IgG HRP Linked Whole Ab, Cytiva NA931-1ML