MilliporeSigma
  • A novel variant in the SLCO2A1 gene in a Chinese patient with chronic gastroenteropathy and primary hypertrophic osteoarthropathy.
A novel variant in the SLCO2A1 gene in a Chinese patient with chronic gastroenteropathy and primary hypertrophic osteoarthropathy.
Orphanet journal of rare diseases (2024-06-12)
Yimin Dai, Miao He, Hui Xu, Bei Tan, Weixun Zhou, Wei Liu, Qiang Wang, Jingyi Huang, Qing Shang, Yaping Liu, Yue Li
ABSTRACT
Chronic enteropathy associated with SLCO2A1 gene (CEAS) results from loss-of-function variants in SLCO2A1, which encodes the prostaglandin transporter (PGT). CEAS follows an autosomal recessive inheritance pattern. To date, approximate 30 pathogenic variants have been reported in CEAS. We performed whole exome sequencing (WES) to screen for potential pathogenic variants in a patient suspected of having CEAS, and confirmed a variant in SLCO2A1 using Sanger sequencing. We established an in vitro minigene model to compare splicing between wild type (WT) and mutant transcripts. Quantitative polymerase chain reaction (qPCR) was used to evaluate SLCO2A1 transcription in the stomach and colon tissues from the patient and a healthy control (HC). The transcripts were further cloned and sequenced. The patient had a novel, homozygous, recessive c.929A > G variant in exon 7 of SLCO2A1, which has not been previously reported in CEAS or PHO. This variant altered splicing, resulting in an exon 7-truncated transcript lacking 16 bases. No normal transcript was detected in the patient's stomach or colon tissue. qPCR also showed significantly decreased SLCO2A1 transcription compared to HC. A previously unreported variant caused defective SLCO2A1 splicing and reduced mRNA levels in a patient with CEAS and PHO. This research enhances understanding of CEAS and PHO pathophysiology and aids genetic counseling and diagnosis.
Sigma-Aldrich
Anti-SLCO2A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution