MilliporeSigma
  • High-throughput screening unveils nitazoxanide as a potent PRRSV inhibitor by targeting NMRAL1.
High-throughput screening unveils nitazoxanide as a potent PRRSV inhibitor by targeting NMRAL1.
Nature communications (2024-06-07)
Zhanding Cui, Jinlong Liu, Chong Xie, Tao Wang, Pu Sun, Jinlong Wang, Jiaoyang Li, Guoxiu Li, Jicheng Qiu, Ying Zhang, Dengliang Li, Ying Sun, Juanbin Yin, Kun Li, Zhixun Zhao, Hong Yuan, Xingwen Bai, Xueqing Ma, Pinghua Li, Yuanfang Fu, Huifang Bao, Dong Li, Qiang Zhang, Zaixin Liu, Yimei Cao, Jing Zhang, Zengjun Lu
ABSTRACT
Porcine Reproductive and Respiratory Syndrome Virus (PRRSV) poses a major threat to the global swine industry, yet effective prevention and control measures remain elusive. This study unveils Nitazoxanide (NTZ) as a potent inhibitor of PRRSV both in vitro and in vivo. Through High-Throughput Screening techniques, 16 potential anti-PRRSV compounds are identified from a library comprising FDA-approved and pharmacopeial drugs. We show that NTZ displays strong efficacy in reducing PRRSV proliferation and transmission in a swine model, alleviating viremia and lung damage. Additionally, Tizoxanide (TIZ), the primary metabolite of NTZ, has been identified as a facilitator of NMRAL1 dimerization. This finding potentially sheds light on the underlying mechanism contributing to TIZ's role in augmenting the sensitivity of the IFN-β pathway. These results indicate the promising potential of NTZ as a repurposed therapeutic agent for Porcine Reproductive and Respiratory Syndrome (PRRS). Additionally, they provide valuable insights into the antiviral mechanisms underlying NTZ's effectiveness.
Sigma-Aldrich
Anti-GATM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution