MilliporeSigma
  • Early developmental deletion of forebrain Ank2 causes seizure-related phenotypes by reshaping the synaptic proteome.
Early developmental deletion of forebrain Ank2 causes seizure-related phenotypes by reshaping the synaptic proteome.
Cell reports (2023-07-10)
Sehyoun Yoon, Marc Dos Santos, Marc P Forrest, Christopher P Pratt, Natalia Khalatyan, Peter J Mohler, Jeffrey N Savas, Peter Penzes
ABSTRACT
Rare genetic variants in ANK2, which encodes ankyrin-B, are associated with neurodevelopmental disorders (NDDs); however, their pathogenesis is poorly understood. We find that mice with prenatal deletion in cortical excitatory neurons and oligodendrocytes (Ank2-/-:Emx1-Cre), but not with adolescent deletion in forebrain excitatory neurons (Ank2-/-:CaMKIIα-Cre), display severe spontaneous seizures, increased mortality, hyperactivity, and social deficits. Calcium imaging of cortical slices from Ank2-/-:Emx1-Cre mice shows increased neuronal calcium event amplitude and frequency, along with network hyperexcitability and hypersynchrony. Quantitative proteomic analysis of cortical synaptic membranes reveals upregulation of dendritic spine plasticity-regulatory proteins and downregulation of intermediate filaments. Characterization of the ankyrin-B interactome identifies interactors associated with autism and epilepsy risk factors and synaptic proteins. The AMPA receptor antagonist, perampanel, restores cortical neuronal activity and partially rescues survival in Ank2-/-:Emx1-Cre mice. Our findings suggest that synaptic proteome alterations resulting from Ank2 deletion impair neuronal activity and synchrony, leading to NDDs-related behavioral impairments.
Sigma-Aldrich
Valproic acid sodium salt, 98%
Sigma-Aldrich
Anti-Glutamate receptor 1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-GluR-2 Antibody, CT, serum, from rabbit
Sigma-Aldrich
Anti-Kalirin Antibody, Upstate®, from rabbit