MilliporeSigma
  • Comparison of the multi-epitope recombinant antigen DIPOL and hydatid fluid for the diagnosis of patients with cystic echinococcosis.
Comparison of the multi-epitope recombinant antigen DIPOL and hydatid fluid for the diagnosis of patients with cystic echinococcosis.
Acta tropica (2021-10-24)
Eylem Akdur Ozturk, Raúl Manzano-Román, Carlos Sánchez-Ovejero, Ayse Caner, Mesude Angın, Cumhur Gunduz, Ülkü Karaman, Nuray Altintas, Halil Bozkaya, Omer Unalp, Zafer Dokumcu, Emre Divarci, Adriano Casulli, Nazmiye Altintas, Mar Siles-Lucas, Aysegul Unver
ABSTRACT
The use of serological tests containing multiple immunodominant antigens rather than single antigens have the potential to improve the diagnostic performance in Cystic Echinococcoses (CE) as a complement tool to clear the inconclusive imaging data. Here, we comparatively evaluated the diagnostic value of Hydatid Fluid (HF) and the recently described recombinant multi-epitope antigen DIPOL in IgG-ELISA in a clinically defined cohort of CE patients. The serum samples from 149 CE patients were collected just before surgical or Percutaneous- Aspiration- Injection- Reaspiration (PAIR) procedures. Additionally, serum samples of patients with other parasitic infections (n=49) and healthy individuals (n=21) were also included in the study as controls. To investigate the association between the genotype of the parasite and DIPOL, cyst materials from 20 CE patients were sequenced. In terms of overall sensitivity, HF was higher than DIPOL (82.55%,78.52%, respectively). However, while the sensitivity of HF was higher than DIPOL in patients with active and transitional cysts (83.3%, 75.4%, respectively), sensitivity of DIPOL in inactive cysts was higher compared to HF (95.6%, 78.3%, respectively). The sensitivity of DIPOL depending on cyst stage was statistically significant (P= 0.041). In terms of specificity, DIPOL was found to be better than HF (97.71%, 91.43%, respectively). By genotyping, the majority of 20 patients showed G1 genotype (80%). All patients harboring G3 and G1/G3 cyst genotypes were positive with both antigens, while 87.5% of patients with G1 genotype were seropositive with HF and 75% with DIPOL. The overall sensitivity and high specificity of DIPOL suggest that this recombinant protein containing immunodominant epitopes is a potential substitute for the HF by serological tests for the diagnosis of CE.
Sigma-Aldrich
Goat Anti-Human IgG Antibody, Alkaline Phosphatase conjugate, Chemicon®, from goat