MilliporeSigma
  • The HDAC6-RNF168 axis regulates H2A/H2A.X ubiquitination to enable double-strand break repair.
The HDAC6-RNF168 axis regulates H2A/H2A.X ubiquitination to enable double-strand break repair.
Nucleic acids research (2023-07-28)
Lingyu Qiu, Wenchao Xu, Xiaopeng Lu, Feng Chen, Yongcan Chen, Yuan Tian, Qian Zhu, Xiangyu Liu, Yongqing Wang, Xin-Hai Pei, Xingzhi Xu, Jun Zhang, Wei-Guo Zhu
ABSTRACT
Histone deacetylase 6 (HDAC6) mediates DNA damage signaling by regulating the mismatch repair and nucleotide excision repair pathways. Whether HDAC6 also mediates DNA double-strand break (DSB) repair is unclear. Here, we report that HDAC6 negatively regulates DSB repair in an enzyme activity-independent manner. In unstressed cells, HDAC6 interacts with H2A/H2A.X to prevent its interaction with the E3 ligase RNF168. Upon sensing DSBs, RNF168 rapidly ubiquitinates HDAC6 at lysine 116, leading to HDAC6 proteasomal degradation and a restored interaction between RNF168 and H2A/H2A.X. H2A/H2A.X is ubiquitinated by RNF168, precipitating the recruitment of DSB repair factors (including 53BP1 and BRCA1) to chromatin and subsequent DNA repair. These findings reveal novel regulatory machinery based on an HDAC6-RNF168 axis that regulates the H2A/H2A.X ubiquitination status. Interfering with this axis might be leveraged to disrupt a key mechanism of cancer cell resistance to genotoxic damage and form a potential therapeutic strategy for cancer.
Sigma-Aldrich
Anti-E3 ubiquitin-protein ligase UHRF1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)