MilliporeSigma
  • Engineering Mycoplasma pneumoniae to bypass the association with Guillain-Barré syndrome.
Engineering Mycoplasma pneumoniae to bypass the association with Guillain-Barré syndrome.
Microbes and infection (2024-04-29)
Alicia Broto, Carlos Piñero-Lambea, Carolina Segura-Morales, Anne P Tio-Gillen, Wendy W J Unger, Raul Burgos, Rocco Mazzolini, Samuel Miravet-Verde, Bart C Jacobs, Josefina Casas, Ruth Huizinga, Maria Lluch-Senar, Luis Serrano
ABSTRACT
A non-pathogenic Mycoplasma pneumoniae-based chassis is leading the development of live biotherapeutic products (LBPs) for respiratory diseases. However, reports connecting Guillain-Barré syndrome (GBS) cases to prior M. pneumoniae infections represent a concern for exploiting such a chassis. Galactolipids, especially galactocerebroside (GalCer), are considered the most likely M. pneumoniae antigens triggering autoimmune responses associated with GBS development. In this work, we generated different strains lacking genes involved in galactolipids biosynthesis. Glycolipid profiling of the strains demonstrated that some mutants show a complete lack of galactolipids. Cross-reactivity assays with sera from GBS patients with prior M. pneumoniae infection showed that certain engineered strains exhibit reduced antibody recognition. However, correlation analyses of these results with the glycolipid profile of the engineered strains suggest that other factors different from GalCer contribute to sera recognition, including total ceramide levels, dihexosylceramide (DHCer), and diglycosyldiacylglycerol (DGDAG). Finally, we discuss the best candidate strains as potential GBS-free Mycoplasma chassis.
Sigma-Aldrich
Anti-Galactocerebroside antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody