MilliporeSigma
  • Preneoplastic cells switch to Warburg metabolism from their inception exposing multiple vulnerabilities for targeted elimination.
Preneoplastic cells switch to Warburg metabolism from their inception exposing multiple vulnerabilities for targeted elimination.
Oncogenesis (2024-01-26)
Henna Myllymäki, Lisa Kelly, Abigail M Elliot, Roderick N Carter, Jeanette Astorga Johansson, Kai Yee Chang, Justyna Cholewa-Waclaw, Nicholas M Morton, Yi Feng
ABSTRACT
Otto Warburg described tumour cells as displaying enhanced aerobic glycolysis whilst maintaining defective oxidative phosphorylation (OXPHOS) for energy production almost 100 years ago [1, 2]. Since then, the 'Warburg effect' has been widely accepted as a key feature of rapidly proliferating cancer cells [3-5]. What is not clear is how early "Warburg metabolism" initiates in cancer and whether changes in energy metabolism might influence tumour progression ab initio. We set out to investigate energy metabolism in the HRASG12V driven preneoplastic cell (PNC) at inception, in a zebrafish skin PNC model. We find that, within 24 h of HRASG12V induction, PNCs upregulate glycolysis and blocking glycolysis reduces PNC proliferation, whilst increasing available glucose enhances PNC proliferation and reduces apoptosis. Impaired OXPHOS accompanies enhanced glycolysis in PNCs, and a mild complex I inhibitor, metformin, selectively suppresses expansion of PNCs. Enhanced mitochondrial fragmentation might be underlining impaired OXPHOS and blocking mitochondrial fragmentation triggers PNC apoptosis. Our data indicate that altered energy metabolism is one of the earliest events upon oncogene activation in somatic cells, which allows a targeted and effective PNC elimination.
Sigma-Aldrich
Mitochondrial Division Inhibitor, mdivi-1, The Mitochondrial Division Inhibitor, mdivi-1, also referenced under CAS 338967-87-6, controls the biological activity of yeast Dnm1 and mammalian Drp1.