MilliporeSigma
  • Wnts acting through canonical and noncanonical signaling pathways exert opposite effects on hippocampal synapse formation.
Wnts acting through canonical and noncanonical signaling pathways exert opposite effects on hippocampal synapse formation.
Neural development (2008-11-07)
Elizabeth K Davis, Yimin Zou, Anirvan Ghosh
ABSTRACT
Wnt proteins comprise a large class of signaling molecules that regulate a variety of developmental processes, including synapse formation. Previous studies have shown Wnts to be involved in both the induction and prevention of synapses in a number of different organisms. However, it is not clear whether the influence of Wnts on synapses is a result of Wnts' behavior in different organisms or differences in the activity of different Wnt ligands. We used in situ hybridization to show that several Wnt ligands (Wnt3, Wnt5a, Wnt7a, and Wnt7b) and their receptors, Frizzled, are expressed in the developing hippocampus during the period of synapse formation in rodents. We used recombinant Wnt protein or Wnt conditioned media to explore the effects of Wnts on synapses in hippocampal cultures. We found that Wnt7a and Wnt7b activate canonical signaling, whereas Wnt5a activates a noncanonical pathway. The activation of the canonical pathway, either through pathway manipulations or through Wnt stimulation, increases presynaptic inputs. In contrast, exposure to Wnt5a, which activates a noncanonical signaling pathway, decreases the number of presynaptic terminals. Our observations suggest that the pro- and antisynaptogenic effects of Wnt proteins are associated with the activation of the canonical and noncanonical Wnt signaling pathways.
Sigma-Aldrich
Laminin from human fibroblasts, cell culture derived, liquid, sterile-filtered
Sigma-Aldrich
Laminin from human placenta, liquid, BioReagent, suitable for cell culture