MilliporeSigma
  • The endogenous brain constituent N-arachidonoyl L-serine is an activator of large conductance Ca2+-activated K+ channels.
The endogenous brain constituent N-arachidonoyl L-serine is an activator of large conductance Ca2+-activated K+ channels.
The Journal of pharmacology and experimental therapeutics (2008-10-17)
Grzegorz Godlewski, László Offertáler, Douglas Osei-Hyiaman, Fong Ming Mo, Judith Harvey-White, Jie Liu, Margaret I Davis, Li Zhang, Raj K Razdan, Garry Milman, Pal Pacher, Partha Mukhopadhyay, David M Lovinger, George Kunos
ABSTRACT
The novel endocannabinoid-like lipid N-arachidonoyl L-serine (ARA-S) causes vasodilation through both endothelium-dependent and -independent mechanisms. We have analyzed the vasorelaxant effect of ARA-S in isolated vascular preparations and its effects on Ca(2+)-activated K(+) currents in human embryonic kidney cells stably transfected with the alpha-subunit of the human, large conductance Ca(+)-activated K(+) (BK(Ca)) channel [human embryonic kidney (HEK) 293hSlo cells]. ARA-S caused relaxation of rat isolated, intact and denuded, small mesenteric arteries preconstricted with (R)-(-)-1-(3-hydroxyphenyl)-2-methylaminoethanol hydrochloride (pEC(50), 5.49 and 5.14, respectively), whereas it caused further contraction of vessels preconstricted with KCl (pEC(50), 5.48 and 4.82, respectively). Vasorelaxation by ARA-S was inhibited by 100 nM iberiotoxin. In human embryonic kidney cells stably transfected with the alpha-subunit of the human BK(Ca) channel cells, ARA-S and its enantiomer, N-arachidonoyl-D-serine, enhanced the whole-cell outward K(+) current with similar potency (pEC(50), 5.63 and 5.32, respectively). The potentiation was not altered by the beta(1) subunit or mediated by ARA-S metabolites, stimulation of known cannabinoid receptors, G proteins, protein kinases, or Ca(2+)-dependent processes; it was lost after patch excision or after membrane cholesterol depletion but was restored after cholesterol reconstitution. BK(Ca) currents were also enhanced by N-arachidonoyl ethanolamide (pEC(50), 5.27) but inhibited by another endocannabinoid, O-arachidonoyl ethanolamine (pIC(50), 6.35), or by the synthetic cannabinoid O-1918 [(-)-1,3-dimethoxy-2-(3-3,4-trans-p-menthadien-(1,8)-yl)-orcinol] (pIC(50), 6.59), which blocks ARA-S-induced vasodilation. We conclude the following. 1) ARA-S directly activates BK(Ca) channels. 2) This interaction does not involve cannabinoid receptors or cytosolic factors but is dependent on the presence of membrane cholesterol. 3) Direct BK(Ca) channel activation probably contributes to the endothelium-independent component of ARA-S-induced mesenteric vasorelaxation. 4) O-1918 is a BK(Ca) channel inhibitor.
Product Description
Supelco
Bradford Reagent, for 0.1-1.4 mg/ml protein
Avanti
N-20:4 L-Serine (ARA-S), N-arachidonoyl L-serine, chloroform
Avanti
23:2 Diyne PE [DC(8,9)PE], 1,2-bis(10,12-tricosadiynoyl)-sn-glycero-3-phosphoethanolamine, powder