MilliporeSigma
  • Dopamine pre-treatment impairs the anti-cancer effect of integrated stress response- and TRAIL pathway-inducing ONC201, ONC206 and ONC212 imipridones in pancreatic, colorectal cancer but not DMG cells.
Dopamine pre-treatment impairs the anti-cancer effect of integrated stress response- and TRAIL pathway-inducing ONC201, ONC206 and ONC212 imipridones in pancreatic, colorectal cancer but not DMG cells.
American journal of cancer research (2024-06-11)
Yiqun Zhang, Nikos Tapinos, Rishi Lulla, Wafik S El-Deiry
ABSTRACT
ONC201 (originally discovered as TRAIL-Inducing Compound #10 or TIC10) and analogue ONC206 have been found to induce an integrated stress response with suggested primary targets and mechanisms involving targeting mitochondrial protein ClpP and antagonism of dopamine receptors D2/3 (DRD2/3). We hypothesized that dopamine, the agonist of DRD2, may counteract ONC201 or ONC206 for DRD2/3 and impair the anti-cancer effect of ONC201 or ONC206, thus protect the tumor cells from the cytotoxic effect of ONC201 or ONC206. We therefore pre-treated cancer cells from different tissue origins including breast cancer, pancreatic cancer, colorectal cancer, and diffuse midline glioma (DMG) with dopamine, followed by treatment of ONC201, ONC206 or ONC212. We observed that 48 hours of pre-treatment with dopamine impaired the cell viability suppression effect of ONC201, ONC206 and ONC212 in pancreatic cancer cells and colorectal cancer cells. We pre-treated multiple cancer cell lines with dopamine for one week followed by ONC201, ONC206, or ONC212 treatment and performed colony assays. Pre-treatment with dopamine impaired the anti-cancer effect of ONC201 or ONC206 in pancreatic cancer and colorectal cancer. Impairment of ONC212 effect by pre-treatment with dopamine was also seen in colony assay for colorectal cancer, but not in pancreatic cancer cells by colony assay. No protection from killing by imipridones was observed with DRD2 agonist sumanirole in tumor cells, or with brain tumor cell lines pretreated with dopamine. Immunoblotting was conducted to investigate whether dopamine pre-treatment impacts signaling pathways reported to be affected by ONC201. The dopamine pre-treatment did not impact changes in ATF4, CHOP, DR5 and ClpX which were reported to be affected by ONC201. The mechanism of impairment of ONC201/206/212 effect caused by dopamine pre-treatment appears to involve upregulation of anti-apoptotic p-Bad, XIAP, FLIP and pAkt. Our results shed light on mechanisms of cancer cell protection by dopamine after imipridone treatment, heterogeneity among different tumor cell types, and suggest that effects of dopamine adaptation on tumor cells may impact on cell survival pathways in ways that may or may not depend on expression of dopamine receptors.
Sigma-Aldrich
Anti-CLPX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid