MilliporeSigma
  • Water from Nitrodi's Spring Induces Dermal Fibroblast and Keratinocyte Activation, Thus Promoting Wound Repair in the Skin: An In Vitro Study.
Water from Nitrodi's Spring Induces Dermal Fibroblast and Keratinocyte Activation, Thus Promoting Wound Repair in the Skin: An In Vitro Study.
International journal of molecular sciences (2023-03-30)
Filomena Napolitano, Loredana Postiglione, Ilaria Mormile, Valentina Barrella, Amato de Paulis, Nunzia Montuori, Francesca Wanda Rossi
ABSTRACT
The Romans knew of Nitrodi's spring on the island of Ischia more than 2000 years ago. Although the health benefits attributed to Nitrodi's water are numerous, the underlying mechanisms are still not understood. In this study, we aim to analyze the physicochemical properties and biological effects of Nitrodi's water on human dermal fibroblasts to determine whether the water exerts in vitro effects that could be relevant to skin wound healing. The results obtained from the study indicate that Nitrodi's water exerts strong promotional effects on dermal fibroblast viability and a significant stimulatory activity on cell migration. Nitrodi's water induces alpha-SMA expression in dermal fibroblasts, thus promoting their transition to myofibroblast-protein ECM deposition. Furthermore, Nitrodi's water reduces intracellular reactive oxygen species (ROS), which play an important role in human skin aging and dermal damage. Unsurprisingly, Nitrodi's water has significant stimulatory effects on the cell proliferation of epidermal keratinocytes and inhibits the basal ROS production but enhances their response to the oxidative stress caused by external stimuli. Our results will contribute to the development of human clinical trials and further in vitro studies to identify inorganic and/or organic compounds responsible for pharmacological effects.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Vitronectin Antibody, clone BV2, clone BV2, Chemicon®, from mouse