MilliporeSigma
  • Slitrk4 is required for the development of inhibitory neurons in the fear memory circuit of the lateral amygdala.
Slitrk4 is required for the development of inhibitory neurons in the fear memory circuit of the lateral amygdala.
Frontiers in molecular neuroscience (2024-05-13)
Yoshifumi Matsumoto, Hideki Miwa, Kei-Ichi Katayama, Arata Watanabe, Kazuyuki Yamada, Takashi Ito, Shinsuke Nakagawa, Jun Aruga
ABSTRACT
The Slitrk family consists of six synaptic adhesion molecules, some of which are associated with neuropsychiatric disorders. In this study, we aimed to investigate the physiological role of Slitrk4 by analyzing Slitrk4 knockout (KO) mice. The Slitrk4 protein was widely detected in the brain and was abundant in the olfactory bulb and amygdala. In a systematic behavioral analysis, male Slitrk4 KO mice exhibited an enhanced fear memory acquisition in a cued test for classical fear conditioning, and social behavior deficits in reciprocal social interaction tests. In an electrophysiological analysis using amygdala slices, Slitrk4 KO mice showed enhanced long-term potentiation in the thalamo-amygdala afferents and reduced feedback inhibition. In the molecular marker analysis of Slitrk4 KO brains, the number of calretinin (CR)-positive interneurons was decreased in the anterior part of the lateral amygdala nuclei at the adult stage. In in vitro experiments for neuronal differentiation, Slitrk4-deficient embryonic stem cells were defective in inducing GABAergic interneurons with an altered response to sonic hedgehog signaling activation that was involved in the generation of GABAergic interneuron subsets. These results indicate that Slitrk4 function is related to the development of inhibitory neurons in the fear memory circuit and would contribute to a better understanding of osttraumatic stress disorder, in which an altered expression of Slitrk4 has been reported.
Sigma-Aldrich
Anti-Somatostatin Antibody, clone YC7, culture supernatant, clone YC7, Chemicon®
Sigma-Aldrich
Anti-Parvalbumin Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Vesicular GABA Transporter (VGAT) Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®