MilliporeSigma
  • Aqueous humor TGFβ and fibrillin-1 in Tsk mice reveal clues to POAG pathogenesis.
Aqueous humor TGFβ and fibrillin-1 in Tsk mice reveal clues to POAG pathogenesis.
Scientific reports (2024-02-13)
James C Tan, MinHee K Ko, Jeong-Im Woo, Kenneth L Lu, Jonathan A Kelber
ABSTRACT
Aqueous humor (AH) and blood levels of transforming growth factor β (TGFβ) are elevated in idiopathic primary open angle glaucoma (POAG) representing a disease biomarker of unclear status and function. Tsk mice display a POAG phenotype and harbor a mutation of fibrillin-1, an important regulator of TGFβ bioavailability. AH TGFβ2 was higher in Tsk than wild-type (WT) mice (by 34%; p = 0.002; ELISA); similarly, AH TGFβ2 was higher in human POAG than controls (2.7-fold; p = 0.00005). As in POAG, TGFβ1 was elevated in Tsk serum (p = 0.01). Fibrillin-1 was detected in AH from POAG subjects and Tsk mice where both had similar levels relative to controls (p = 0.45). 350 kDa immunoblot bands representing WT full-length fibrillin-1 were present in human and mouse AH. A 418 kDa band representing mutant full-length fibrillin-1 was present only in Tsk mice. Lower molecular weight fibrillin-1 antibody-reactive bands were present in similar patterns in humans and mice. Certain bands (130 and 32 kDa) were elevated only in human POAG and Tsk mice (p ≤ 0.04 relative to controls) indicating discrete isoforms relevant to disease. In addition to sharing a phenotype, Tsk mice and human POAG subjects had common TGFβ and fibrillin-1 features in AH and also blood that are pertinent to understanding glaucoma pathogenesis.
Sigma-Aldrich
Anti-Fibrillin-1 Antibody, NT, clone 26, clone 26, Chemicon®, from mouse