MilliporeSigma
  • Aberrant Mucin Expression Profiles Associate With Pediatric Inflammatory Bowel Disease Presentation and Activity.
Aberrant Mucin Expression Profiles Associate With Pediatric Inflammatory Bowel Disease Presentation and Activity.
Inflammatory bowel diseases (2022-10-15)
Tom Breugelmans, Wout Arras, Lauren-Emma Boen, Eliah Borms, Lisa Kamperdijk, Joris De Man, Els Van de Vijver, Ann Van Gils, Benedicte Y De Winter, Nicolette Moes, Annemieke Smet
ABSTRACT
Intestinal mucosal healing is nowadays preferred as the therapeutic endpoint in inflammatory bowel disease (IBD), but objective measurements at the molecular level are lacking. Because dysregulated mucin expression is suggested to be involved in mucosal barrier dysfunction in IBD, we investigated mucin expression in association with barrier mediators and clinical characteristics in colonic tissue of a pediatric IBD population. In this cross-sectional monocentric study, we quantified messenger RNA (mRNA) expression of mucins, intercellular junctions, and cell polarity complexes in inflamed and noninflamed colonic biopsies from pediatric IBD (n = 29) and non-IBD (n = 15) patients. We then validated mucin expression at protein level and correlated mucin mRNA expression with expression of barrier mediators and clinical data. The expression of MUC1, MUC3A, MUC4, and MUC13 was increased in the inflamed colon of pediatric IBD patients compared with the noninflamed colon of non-IBD control subjects. Especially MUC13 mRNA expression associated with the expression of barrier mediators, including CDH1, OCLN, and TJP2. MUC1 and MUC3B mRNA expression in combination with calprotectin levels most accurately discriminated IBD patients from non-IBD control subjects (90.6% area under the receiver-operating characteristic curve [AUCROC], 92.0% sensitivity, 73.7% specificity), whereas aberrant mRNA expression of MUC1, MUC3A, MUC4, and MUC13 was distinctive for ulcerative colitis and of MUC3B for Crohn's disease. Furthermore, expression of MUC3A, MUC3B, and MUC4 correlated with clinical disease activity (ie, Pediatric Ulcerative Colitis Activity Index and Pediatric Crohn's Disease Activity Index), and of MUC1, MUC2, MUC4, and MUC13 with endoscopic colitis severity in ulcerative colitis patients. Colonic mucin expression is disturbed in pediatric IBD patients and associates with disease activity and presentation, suggesting its use as molecular marker to aid in disease diagnosis and management.
Sigma-Aldrich
Anti-MUC13 Antibody, clone 2E11.1, clone 2E11.1, from mouse