MilliporeSigma
  • mTORC2-driven chromatin cGAS mediates chemoresistance through epigenetic reprogramming in colorectal cancer.
mTORC2-driven chromatin cGAS mediates chemoresistance through epigenetic reprogramming in colorectal cancer.
Nature cell biology (2024-07-31)
Guoqing Lv, Qian Wang, Lin Lin, Qiao Ye, Xi Li, Qian Zhou, Xiangzhen Kong, Hongxia Deng, Fuping You, Hebing Chen, Song Wu, Lin Yuan
ABSTRACT
Cyclic GMP-AMP synthase (cGAS), a cytosolic DNA sensor that initiates a STING-dependent innate immune response, binds tightly to chromatin, where its catalytic activity is inhibited; however, mechanisms underlying cGAS recruitment to chromatin and functions of chromatin-bound cGAS (ccGAS) remain unclear. Here we show that mTORC2-mediated phosphorylation of human cGAS serine 37 promotes its chromatin localization in colorectal cancer cells, regulating cell growth and drug resistance independently of STING. We discovered that ccGAS recruits the SWI/SNF complex at specific chromatin regions, modifying expression of genes linked to glutaminolysis and DNA replication. Although ccGAS depletion inhibited cell growth, it induced chemoresistance to fluorouracil treatment in vitro and in vivo. Moreover, blocking kidney-type glutaminase, a downstream ccGAS target, overcame chemoresistance caused by ccGAS loss. Thus, ccGAS coordinates colorectal cancer plasticity and acquired chemoresistance through epigenetic patterning. Targeting both mTORC2-ccGAS and glutaminase provides a promising strategy to eliminate quiescent resistant cancer cells.
Product Description
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Anti-SMARCA4 Antibody, clone 20C3.2, clone 20C3.2, from mouse
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-MCM5 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution