MilliporeSigma
  • Annexin A1 binds PDZ and LIM domain 7 to inhibit adipogenesis and prevent obesity.
Annexin A1 binds PDZ and LIM domain 7 to inhibit adipogenesis and prevent obesity.
Signal transduction and targeted therapy (2024-08-23)
Lu Fang, Changjie Liu, Zong-Zhe Jiang, Mengxiao Wang, Kang Geng, Yangkai Xu, Yujie Zhu, Yiwen Fu, Jing Xue, Wenxin Shan, Qi Zhang, Jie Chen, Jiahong Chen, Mingming Zhao, Yuxuan Guo, K W Michael Siu, Y Eugene Chen, Yong Xu, Donghui Liu, Lemin Zheng
ABSTRACT
Obesity is a global issue that warrants the identification of more effective therapeutic targets and a better understanding of the pivotal molecular pathogenesis. Annexin A1 (ANXA1) is known to inhibit phospholipase A2, exhibiting anti-inflammatory activity. However, the specific effects of ANXA1 in obesity and the underlying mechanisms of action remain unclear. Our study reveals that ANXA1 levels are elevated in the adipose tissue of individuals with obesity. Whole-body or adipocyte-specific ANXA1 deletion aggravates obesity and metabolic disorders. ANXA1 levels are higher in stromal vascular fractions (SVFs) than in mature adipocytes. Further investigation into the role of ANXA1 in SVFs reveals that ANXA1 overexpression induces lower numbers of mature adipocytes, while ANXA1-knockout SVFs exhibit the opposite effect. This suggests that ANXA1 plays an important role in adipogenesis. Mechanistically, ANXA1 competes with MYC binding protein 2 (MYCBP2) for interaction with PDZ and LIM domain 7 (PDLIM7). This exposes the MYCBP2-binding site, allowing it to bind more readily to the SMAD family member 4 (SMAD4) and promoting its ubiquitination and degradation. SMAD4 degradation downregulates peroxisome proliferator-activated receptor gamma (PPARγ) transcription and reduces adipogenesis. Treatment with Ac2-26, an active peptide derived from ANXA1, inhibits both adipogenesis and obesity through the mechanism. In conclusion, the molecular mechanism of ANXA1 inhibiting adipogenesis was first uncovered in our study, which is a potential target for obesity prevention and treatment.
Sigma-Aldrich
Anti-MYCBP2 Antibody, clone PM-8A9, clone PM-8A9, from mouse