MilliporeSigma
  • Role of Glutamatergic Projections from Lateral Habenula to Ventral Tegmental Area in Inflammatory Pain-Related Spatial Working Memory Deficits.
Role of Glutamatergic Projections from Lateral Habenula to Ventral Tegmental Area in Inflammatory Pain-Related Spatial Working Memory Deficits.
Biomedicines (2023-03-30)
Mobina Alemi, Ana Raquel Pereira, Mariana Cerqueira-Nunes, Clara Monteiro, Vasco Galhardo, Helder Cardoso-Cruz
ABSTRACT
The lateral habenula (LHb) and the ventral tegmental area (VTA), which form interconnected circuits, have important roles in the crucial control of sensory and cognitive motifs. Signaling in the LHb-VTA pathway can be exacerbated during pain conditions by a hyperactivity of LHb glutamatergic neurons to inhibit local VTA DAergic cells. However, it is still unclear whether and how this circuit is endogenously engaged in pain-related cognitive dysfunctions. To answer this question, we modulated this pathway by expressing halorhodopsin in LHb neurons of adult male rats, and then selectively inhibited the axonal projections from these neurons to the VTA during a working memory (WM) task. Behavioral performance was assessed after the onset of an inflammatory pain model. We evaluated the impact of the inflammatory pain in the VTA synapses by performing immunohistochemical characterization of specific markers for GABAergic (GAD65/67) and dopaminergic neurons (dopamine transporter (DAT), dopamine D2 receptor (D2r) and tyrosine hydroxylase (TH)). Our results revealed that inhibition of LHb terminals in the VTA during the WM delay-period elicits a partial recovery of the performance of pain animals (in higher complexity challenges); this performance was not accompanied by a reduction of nociceptive responses. Finally, we found evidence that the pain-affected animals exhibit VTA structural changes, namely with an upregulation of GAD65/67, and a downregulation of DAT and D2r. These results demonstrate a role of LHb neurons and highlight their responsibility in the stability of the local VTA network, which regulates signaling in frontal areas necessary to support WM processes.
Sigma-Aldrich
Anti-Dopamine D2 receptor (DRD2) Antibody, clone 2B9, clone 3D9, from mouse
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®