MilliporeSigma
  • Cortical somatostatin long-range projection neurons and interneurons exhibit divergent developmental trajectories.
Cortical somatostatin long-range projection neurons and interneurons exhibit divergent developmental trajectories.
Neuron (2023-12-13)
Josephine Fisher, Marieke Verhagen, Zhen Long, Monika Moissidis, Yiming Yan, Chenyi He, Jingyu Wang, Elia Micoli, Clara Milían Alastruey, Rani Moors, Oscar Marín, Da Mi, Lynette Lim
ABSTRACT
The mammalian cerebral cortex contains an extraordinary diversity of cell types that emerge by implementing different developmental programs. Delineating when and how cellular diversification occurs is particularly challenging for cortical inhibitory neurons because they represent a small proportion of all cortical cells and have a protracted development. Here, we combine single-cell RNA sequencing and spatial transcriptomics to characterize the emergence of neuronal diversity among somatostatin-expressing (SST+) cells in mice. We found that SST+ inhibitory neurons segregate during embryonic stages into long-range projection (LRP) neurons and two types of interneurons, Martinotti cells and non-Martinotti cells, following distinct developmental trajectories. Two main subtypes of LRP neurons and several subtypes of interneurons are readily distinguishable in the embryo, although interneuron diversity is likely refined during early postnatal life. Our results suggest that the timing for cellular diversification is unique for different subtypes of SST+ neurons and particularly divergent for LRP neurons and interneurons.
Sigma-Aldrich
Anti-NNOS antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-MEIS2 antibody produced in mouse, clone 1H4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Nitric Oxide Synthase I Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Somatostatin Antibody, clone YC7, culture supernatant, clone YC7, Chemicon®