MilliporeSigma
  • Tau in the brain interstitial fluid is fragmented and seeding-competent.
Tau in the brain interstitial fluid is fragmented and seeding-competent.
Neurobiology of aging (2021-10-17)
Erica Barini, Gudrun Plotzky, Yulia Mordashova, Jonas Hoppe, Esther Rodriguez-Correa, Sonja Julier, Florie LePrieult, Ina Mairhofer, Mario Mezler, Sandra Biesinger, Miroslav Cik, Marcus W Meinhardt, Ebru Ercan-Herbst, Dagmar E Ehrnhoefer, Andreas Striebinger, Karen Bodie, Corinna Klein, Laura Gasparini, Kerstin Schlegel
ABSTRACT
In Alzheimer disease, Tau pathology is thought to propagate from cell to cell throughout interconnected brain areas. However, the forms of Tau released into the brain interstitial fluid (ISF) in vivo during the development of Tauopathy and their pathological relevance remain unclear. Combining in vivo microdialysis and biochemical analysis, we find that in Tau transgenic mice, human Tau (hTau) present in brain ISF is truncated and comprises at least 10 distinct fragments spanning the entire Tau protein. The fragmentation pattern is similar across different Tau transgenic models, pathological stages and brain areas. ISF hTau concentration decreases during Tauopathy progression, while its phosphorylation increases. ISF from mice with established Tauopathy induces Tau aggregation in HEK293-Tau biosensor cells. Notably, immunodepletion of ISF phosphorylated Tau, but not Tau fragments, significantly reduces its ability to seed Tau aggregation and only a fraction of Tau, separated by ultracentrifugation, is seeding-competent. These results indicate that ISF seeding competence is driven by a small subset of Tau, which potentially contribute to the propagation of Tau pathology.
Sigma-Aldrich
IOX1, ≥97% (HPLC)
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.