MilliporeSigma
  • Histological Analysis of Gonadal Ridge Development and Sex Differentiation of Gonads in Three Gecko Species.
Histological Analysis of Gonadal Ridge Development and Sex Differentiation of Gonads in Three Gecko Species.
Biology (2024-01-22)
Izabela Rams-Pociecha, Paulina C Mizia, Rafal P Piprek
ABSTRACT
Reptiles constitute a highly diverse group of vertebrates, with their evolutionary lineages having diverged relatively early. The types of sex determination exemplify the diversity of reptiles; however, there are limited data regarding the gonadal development in squamate reptiles. Geckos constitute a group that is increasingly used in research and that serves as a potential reptilian model organism. The aim of this study was to trace the changes in the structure of developing gonads in the embryos of three gecko species: the crested gecko, leopard gecko, and mourning gecko. These species represent different families of the Gekkota infraorder and exhibit different types of sex determination. Gonadal development was examined from the formation of the earliest gonadal ridges through the development of undifferentiated gonadal structures, sex differentiation of gonads, and the formation of testicular and ovarian structures. The study showed that the gonadal primordia of these three gecko species formed on the most dorsally located surface of the dorsal mesentery, and both the coelomic epithelium and the nephric mesenchyme contributed to their development. As in other reptile species, primordial germ cells settled in the gonadal ridges, and the undifferentiated gonad was composed of a cortex and a medulla. Ovarian differentiation started with the thickening of the gonadal cortex and proliferation of germ cells in this region. A characteristic feature of the developing gecko ovaries was the thickened crescent-shaped cortex on the medial and ventral surfaces of the ovaries. The ovarian medulla also grew and exhibited diverse tendencies to form cords. In the leopard gecko, advanced cord-like structures with lumens were observed in the ovaries, which were not seen in the crested gecko. Testicular differentiation was characterized by cortical thinning and the disappearance of germ cells in this region. In the medulla, the development of distinct cords with early lumen formation was noted. A characteristic feature of embryonic gonads was their growth in a horizontal plane. In this study, gonadal development was characterized by several features that are shared by geckos and other reptiles, along with features that are specific only to geckos.
Sigma-Aldrich
Anti-PCNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution