MilliporeSigma
  • Clinical significance of L1CAM expression in metastatic tubo-ovarian high-grade serous carcinoma.
Clinical significance of L1CAM expression in metastatic tubo-ovarian high-grade serous carcinoma.
Gynecologic oncology (2023-07-22)
Margarida Varela Dos Santos, Arild Holth, Kristina Lindemann, Anne Cathrine Staff, Ben Davidson
ABSTRACT
To analyze the expression and prognostic role of L1CAM in tubo-ovarian high-grade serous carcinoma (HGSC). L1CAM protein expression by immunohistochemistry was analyzed in 644 HGSC (413 effusions, 231 surgical specimens). Expression was analyzed for association with clinicopathologic parameters and survival. L1CAM protein expression was found in 401/413 (97%) effusions and 209/231 (90%) surgical specimens, with significantly higher staining extent in effusions (p < 0.001). L1CAM protein expression in effusions was unrelated to clinicopathologic parameters (p > 0.05). In surgical specimens, higher L1CAM expression was significantly related to primary (intrinsic) chemoresistance (p = 0.017). High (>25%) L1CAM expression in HGSC effusions (p = 0.02), older patient age (p = 0.013), FIGO stage IV disease (p < 0.001) and larger residual disease volume (p = 0.001) were significantly associated with shorter overall survival (OS) in univariate analysis. In Cox multivariate analysis, only FIGO stage (p = 0.001) and residual disease volume (p = 0.003) were independent prognosticators of OS. L1CAM expression in effusions was unrelated to progression-free survival (PFS). There was no association between L1CAM expression in surgical specimens and survival. L1CAM is overexpressed in HGSC effusions compared to surgical specimens. Its overexpression in effusions is significantly associated with shorter OS, but not independently of established prognostic factors such as FIGO stage and residual disease volume.
Sigma-Aldrich
IgG1, Kappa from murine myeloma, clone MOPC 31C, purified immunoglobulin, buffered aqueous solution