MilliporeSigma
  • Exacerbated Activation of the NLRP3 Inflammasome in the Placentas from Women Who Developed Chronic Venous Disease during Pregnancy.
Exacerbated Activation of the NLRP3 Inflammasome in the Placentas from Women Who Developed Chronic Venous Disease during Pregnancy.
International journal of molecular sciences (2024-05-25)
María Asunción Sánchez-Gil, Oscar Fraile-Martinez, Cielo García-Montero, Diego De Leon-Oliva, Diego Liviu Boaru, Patricia De Castro-Martinez, Adrían Camacho-Alcázar, Juan A De León-Luis, Coral Bravo, Raúl Díaz-Pedrero, Laura López-Gonzalez, Julia Bujan, María J Cancelo, Melchor Álvarez-Mon, Natalio García-Honduvilla, Miguel A Saez, Miguel A Ortega
ABSTRACT
Chronic venous disease (CVD) comprises a spectrum of morphofunctional disorders affecting the venous system, affecting approximately 1 in 3 women during gestation. Emerging evidence highlights diverse maternofetal implications stemming from CVD, particularly impacting the placenta. While systemic inflammation has been associated with pregnancy-related CVD, preliminary findings suggest a potential link between this condition and exacerbated inflammation in the placental tissue. Inflammasomes are major orchestrators of immune responses and inflammation in different organs and systems. Notwithstanding the relevance of inflammasomes, specifically the NLRP3 (nucleotide-binding domain, leucine-rich-containing family, pyrin domain-containing-3)- which has been demonstrated in the placentas of women with different obstetric complications, the precise involvement of this component in the placentas of women with CVD remains to be explored. This study employs immunohistochemistry and real-time PCR (RT-qPCR) to examine the gene and protein expression of key components in both canonical and non-canonical pathways of the NLRP3 inflammasome (NLRP3, ASC-apoptosis-associated speck-like protein containing a C-terminal caspase recruitment domain-caspase 1, caspase 5, caspase 8, and interleukin 1β) within the placental tissue of women affected by CVD. Our findings reveal a substantial upregulation of these components in CVD-affected placentas, indicating a potential pathophysiological role of the NLRP3 inflammasome in the development of this condition. Subsequent investigations should focus on assessing translational interventions addressing this dysregulation in affected patient populations.
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture