MilliporeSigma
  • CGRP Released by Corneal Sensory Nerve Maintains Tear Secretion of the Lacrimal Gland.
CGRP Released by Corneal Sensory Nerve Maintains Tear Secretion of the Lacrimal Gland.
Investigative ophthalmology & visual science (2024-04-18)
Linyan Ma, Lingling Yang, Xiaolei Wang, Leilei Zhao, Xiaofei Bai, Xia Qi, Qing Chen, Ya Li, Qingjun Zhou
ABSTRACT
This study aims to elucidate the calcitonin gene-related peptide (CGRP) mediation and primary mechanism of corneal sensory nerves on tear production of the lacrimal gland. Mouse corneal denervation models were constructed through surgical axotomy, pharmacologic treatment with capsaicin or resiniferatoxin, and Trpv1-Cre/DTR mice with diphtheria toxin injection. The capsaicin-treated mice received subconjunctival injection of CGRP or substance P, while the normal C57BL/6J mice were administered with CGRP receptor antagonist BIBN-4096. Furthermore, double immunostaining of c-FOS+ and choline acetyltransferase was used to evaluate the activation of the superior salivatory nucleus (SSN). Mouse lacrimal glands were collected for transcriptomic sequencing and subsequent RNA and protein expression analysis. The corneal denervated mice exhibited a significant reduction in corneal sensitivity and tear secretion. In capsaicin-treated mice, tear secretion decreased to 2.5 ± 0.5 mm compared to 6.3 ± 0.9 mm in control mice (P < 0.0001). However, exogenous administration of CGRP in capsaicin-treated mice increased tear secretion from 2.6 ± 0.5 mm to 4.5 ± 0.5 mm (P = 0.0009), while BIBN-4096 treatment reduced tear secretion to 3.4 ± 0.5 mm when compared to 7.3 ± 0.7 mm in control mice (P = 0.0022). Furthermore, c-FOS+ cell number in the SSN increased by twofold (P = 0.0168) after CGRP administration compared with capsaicin-treated mice. In addition, the expressions of CCNA2, Ki67, PCNA, and CDK1 in acinar cells of the lacrimal gland were impaired by corneal denervation and alleviated by CGRP administration. CGRP released by corneal sensory nerves mediates tear secretion of the lacrimal gland, providing a new strategy for improving tear secretion in patients with neurotrophic keratitis.
Sigma-Aldrich
Monoclonal Anti-CHAT antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL3173, purified immunoglobulin, buffered aqueous glycerol solution
Pricing and availability is not currently available.