MilliporeSigma
  • TGF-beta has paradoxical and context dependent effects on proliferation and anoikis in human colorectal cancer cell lines.
TGF-beta has paradoxical and context dependent effects on proliferation and anoikis in human colorectal cancer cell lines.
Growth factors (Chur, Switzerland) (2008-07-25)
Nina M Muñoz, Ji Yeon Baek, William M Grady
ABSTRACT
Transforming growth factor-beta (TGF-beta) is a pluripotent cytokine that can have both tumor suppressing and tumor promoting effects on epithelial cells. It is unclear what determines when TGF-beta and its signaling pathway act predominantly as a tumor suppressor pathway or as a tumor-promoter pathway and whether TGF-beta can have both classes of effects concurrently on a cell. We investigated the effect of TGF-beta on anoikis in colorectal cancer cell lines sensitive to TGF-beta-mediated growth inhibition to determine if the context of the cells could be one of the factors that would affect whether TGF-beta exerts tumor suppressor or oncogene activity on colon cancer cells. We observed variable effects of TGF-beta on anoikis in these cell lines, even though they all are growth-inhibited by TGF-beta. Thus, we show that TGF-beta has variable effects on anoikis in colon cancer cell lines that likely reflects the effects of concurrent gene mutations in the cancer cells and the activation state of the signaling pathways controlled by these genes.
Sigma-Aldrich
McCoy′s 5A Medium, Modified, with L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
McCoy′s 5A Medium, Modified, with L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
McCoy′s 5A Medium, Modified, with sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)