MilliporeSigma
  • RUVBL1 in Clear-Cell Renal Cell Carcinoma: Unraveling Prognostic Significance and Correlation with HIF1A.
RUVBL1 in Clear-Cell Renal Cell Carcinoma: Unraveling Prognostic Significance and Correlation with HIF1A.
Cancers (2024-04-13)
Justyna Durślewicz, Aleksandra Maria Wybierała, Sara Szczepanek, Paulina Antosik, Damian Jaworski, Dariusz Grzanka
ABSTRACT
This study investigates the roles of RUVBL1 and HIF1A in ccRCC development and explores their clinical significance as prognostic biomarkers. mRNA and protein expressions were analyzed using TCGA data and an institutional tissue cohort, respectively. Correlations with clinicopathological parameters and patient outcomes were assessed. TCGA data revealed significantly elevated RUVBL1 mRNA expression in ccRCC tissues, associated with advanced histological grade, T stage, lymph node metastasis, and clinical stage. High RUVBL1 mRNA expression correlated with inferior overall survival and served as an adverse prognostic factor. Similarly, HIF1A mRNA expression was significantly higher in ccRCC tissues, correlating with worse overall survival and acting as an adverse prognostic factor for treatment outcomes. Simultaneous evaluation of RUVBL1 and HIF1A mRNA expression demonstrated enhanced prognostic capacity, surpassing the predictive power of individual markers. Immunohistochemical staining confirmed substantial upregulation of both RUVBL1 and HIF-1α proteins in ccRCC tissues. Furthermore, high expression of both RUVBL1 and HIF-1α proteins was significantly associated with shorter patient survival time. Our findings underscore the significance of RUVBL1 and HIF-1α as potential prognostic markers in ccRCC, paving the way for further research to translate these insights into clinically relevant applications.
Sigma-Aldrich
Anti-RUVBL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-Hypoxia Inducible Factor 1 α Antibody, clone H1α67, clone H1alpha67, Chemicon®, from mouse