MilliporeSigma
  • A comprehensive Drosophila resource to identify key functional interactions between SARS-CoV-2 factors and host proteins.
A comprehensive Drosophila resource to identify key functional interactions between SARS-CoV-2 factors and host proteins.
Cell reports (2023-07-22)
Annabel Guichard, Shenzhao Lu, Oguz Kanca, Daniel Bressan, Yan Huang, Mengqi Ma, Sara Sanz Juste, Jonathan C Andrews, Kristy L Jay, Marketta Sneider, Ruth Schwartz, Mei-Chu Huang, Danqing Bei, Hongling Pan, Liwen Ma, Wen-Wen Lin, Ankush Auradkar, Pranjali Bhagwat, Soo Park, Kenneth H Wan, Takashi Ohsako, Toshiyuki Takano-Shimizu, Susan E Celniker, Michael F Wangler, Shinya Yamamoto, Hugo J Bellen, Ethan Bier
ABSTRACT
Development of effective therapies against SARS-CoV-2 infections relies on mechanistic knowledge of virus-host interface. Abundant physical interactions between viral and host proteins have been identified, but few have been functionally characterized. Harnessing the power of fly genetics, we develop a comprehensive Drosophila COVID-19 resource (DCR) consisting of publicly available strains for conditional tissue-specific expression of all SARS-CoV-2 encoded proteins, UAS-human cDNA transgenic lines encoding established host-viral interacting factors, and GAL4 insertion lines disrupting fly homologs of SARS-CoV-2 human interacting proteins. We demonstrate the utility of the DCR to functionally assess SARS-CoV-2 genes and candidate human binding partners. We show that NSP8 engages in strong genetic interactions with several human candidates, most prominently with the ATE1 arginyltransferase to induce actin arginylation and cytoskeletal disorganization, and that two ATE1 inhibitors can reverse NSP8 phenotypes. The DCR enables parallel global-scale functional analysis of SARS-CoV-2 components in a prime genetic model system.
Sigma-Aldrich
Anti-beta Actin Antibody, arginylated (N-terminal), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®