MilliporeSigma
  • Apolipoprotein E2 (Lys146-->Gln) causes hypertriglyceridemia due to an apolipoprotein E variant-specific inhibition of lipolysis of very low density lipoproteins-triglycerides.
Apolipoprotein E2 (Lys146-->Gln) causes hypertriglyceridemia due to an apolipoprotein E variant-specific inhibition of lipolysis of very low density lipoproteins-triglycerides.
Arteriosclerosis, thrombosis, and vascular biology (2000-07-15)
F de Beer, K W van Dijk, M C Jong, L C van Vark, A van der Zee, M H Hofker, F J Fallaux, R C Hoeben, A H Smelt, L M Havekes
ABSTRACT
The apolipoprotein E2 (Lys146-->Gln) variant is associated with a dominant form of familial dysbetalipoproteinemia. Heterozygous carriers of this variant have elevated levels of plasma triglycerides, cholesterol, and apolipoprotein E (apoE). It was hypothesized that the high amounts of triglycerides in the very low density lipoprotein (VLDL) fraction are due to a disturbed lipolysis of VLDL. To test this hypothesis, apoE knockout mice were injected with an adenovirus containing the human APOE*2 (Lys146-->Gln) gene, Ad-E2(146), under the control of the cytomegalovirus promoter. ApoE knockout mice injected with an adenovirus vector encoding human apoE3 (Ad-E3) were used as controls. Five days after adenovirus injection, plasma cholesterol levels of mice injected with a high dose of Ad-E2(146) (2x10(9) plaque-forming units) were not changed compared with preinjection levels, whereas in the group who received a low dose of Ad-E2(146) (5x10(8) plaque-forming units) and in the groups injected with a low or a high dose of Ad-E3, plasma cholesterol levels were decreased 5-, 6-, and 12-fold, respectively. Plasma triglycerides were not affected in mice injected with Ad-E3. In contrast, a 7-fold increase in plasma triglycerides was observed in mice injected with the low dose of Ad-E2(146) compared with mice injected with Ad-E3. Injection with the high dose of Ad-E2(146) resulted in a dramatic increase of plasma triglycerides (50-fold compared with Ad-E3 injection). In vitro lipolysis experiments showed that the lipolysis rate of VLDLs containing normal amounts of apoE2 (Lys146-->Gln) was decreased by 54% compared with that of VLDLs containing comparable amounts of apoE3. The in vivo VLDL-triglyceride production rate of Ad-E2(146)-injected mice was not significantly different from that of Ad-E3-injected mice. These results demonstrate that expression of apoE2 (Lys146-->Gln) causes hypertriglyceridemia due to an apoE variant-specific inhibition of the hydrolysis of VLDL-triglycerides.
Sigma-Aldrich
Lipoprotein Lipase from bovine milk, ammonium sulfate suspension, ≥2,000 units/mg protein (BCA)