MilliporeSigma
  • Two distinct gut microbial pathways contribute to meta-organismal production of phenylacetylglutamine with links to cardiovascular disease.
Two distinct gut microbial pathways contribute to meta-organismal production of phenylacetylglutamine with links to cardiovascular disease.
Cell host & microbe (2022-12-23)
Yijun Zhu, Mohammed Dwidar, Ina Nemet, Jennifer A Buffa, Naseer Sangwan, Xinmin S Li, James T Anderson, Kymberleigh A Romano, Xiaoming Fu, Masanori Funabashi, Zeneng Wang, Pooja Keranahalli, Shawna Battle, Aaron N Tittle, Adeline M Hajjar, Valentin Gogonea, Michael A Fischbach, Joseph A DiDonato, Stanley L Hazen
ABSTRACT
Recent studies show gut microbiota-dependent metabolism of dietary phenylalanine into phenylacetic acid (PAA) is critical in phenylacetylglutamine (PAGln) production, a metabolite linked to atherosclerotic cardiovascular disease (ASCVD). Accordingly, microbial enzymes involved in this transformation are of interest. Using genetic manipulation in selected microbes and monocolonization experiments in gnotobiotic mice, we identify two distinct gut microbial pathways for PAA formation; one is catalyzed by phenylpyruvate:ferredoxin oxidoreductase (PPFOR) and the other by phenylpyruvate decarboxylase (PPDC). PPFOR and PPDC play key roles in gut bacterial PAA production via oxidative and non-oxidative phenylpyruvate decarboxylation, respectively. Metagenomic analyses revealed a significantly higher abundance of both pathways in gut microbiomes of ASCVD patients compared with controls. The present studies show a role for these two divergent microbial catalytic strategies in the meta-organismal production of PAGln. Given the numerous links between PAGln and ASCVD, these findings will assist future efforts to therapeutically target PAGln formation in vivo.
Sigma-Aldrich
Nitazoxanide, ≥98% (HPLC)