MilliporeSigma
  • Genetic variants of phospholipase C-γ2 alter the phenotype and function of microglia and confer differential risk for Alzheimer's disease.
Genetic variants of phospholipase C-γ2 alter the phenotype and function of microglia and confer differential risk for Alzheimer's disease.
Immunity (2023-09-03)
Andy P Tsai, Chuanpeng Dong, Peter Bor-Chian Lin, Adrian L Oblak, Gonzalo Viana Di Prisco, Nian Wang, Nicole Hajicek, Adam J Carr, Emma K Lendy, Oliver Hahn, Micaiah Atkins, Aulden G Foltz, Jheel Patel, Guixiang Xu, Miguel Moutinho, John Sondek, Qisheng Zhang, Andrew D Mesecar, Yunlong Liu, Brady K Atwood, Tony Wyss-Coray, Kwangsik Nho, Stephanie J Bissel, Bruce T Lamb, Gary E Landreth
ABSTRACT
Genetic association studies have demonstrated the critical involvement of the microglial immune response in Alzheimer's disease (AD) pathogenesis. Phospholipase C-gamma-2 (PLCG2) is selectively expressed by microglia and functions in many immune receptor signaling pathways. In AD, PLCG2 is induced uniquely in plaque-associated microglia. A genetic variant of PLCG2, PLCG2P522R, is a mild hypermorph that attenuates AD risk. Here, we identified a loss-of-function PLCG2 variant, PLCG2M28L, that confers an increased AD risk. PLCG2P522R attenuated disease in an amyloidogenic murine AD model, whereas PLCG2M28L exacerbated the plaque burden associated with altered phagocytosis and Aβ clearance. The variants bidirectionally modulated disease pathology by inducing distinct transcriptional programs that identified microglial subpopulations associated with protective or detrimental phenotypes. These findings identify PLCG2M28L as a potential AD risk variant and demonstrate that PLCG2 variants can differentially orchestrate microglial responses in AD pathogenesis that can be therapeutically targeted.
Sigma-Aldrich
X-34, ≥90% (HPLC)
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, Alexa Fluor488 Conjugate | MABN50A4, clone 211F1.1, from mouse, ALEXA FLUOR 488