MilliporeSigma
  • IFI16 phase separation via multi-phosphorylation drives innate immune signaling.
IFI16 phase separation via multi-phosphorylation drives innate immune signaling.
Nucleic acids research (2023-06-07)
Dawei Liu, Krystal K Lum, Nicholas Treen, Corazón T Núñez, Jinhang Yang, Timothy R Howard, Michael Levine, Ileana M Cristea
ABSTRACT
The interferon inducible protein 16 (IFI16) is a prominent sensor of nuclear pathogenic DNA, initiating innate immune signaling and suppressing viral transcription. However, little is known about mechanisms that initiate IFI16 antiviral functions or its regulation within the host DNA-filled nucleus. Here, we provide in vitro and in vivo evidence to establish that IFI16 undergoes liquid-liquid phase separation (LLPS) nucleated by DNA. IFI16 binding to viral DNA initiates LLPS and induction of cytokines during herpes simplex virus type 1 (HSV-1) infection. Multiple phosphorylation sites within an intrinsically disordered region (IDR) function combinatorially to activate IFI16 LLPS, facilitating filamentation. Regulated by CDK2 and GSK3β, IDR phosphorylation provides a toggle between active and inactive IFI16 and the decoupling of IFI16-mediated cytokine expression from repression of viral transcription. These findings show how IFI16 switch-like phase transitions are achieved with temporal resolution for immune signaling and, more broadly, the multi-layered regulation of nuclear DNA sensors.
Sigma-Aldrich
Monoclonal Anti-IFI16 antibody produced in mouse, clone 2E3, purified immunoglobulin, buffered aqueous solution
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture